Stampa…

4-(Trifluoromethyl)cyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 4-(Trifluoromethyl)cyclohexane-1-carboxylic acid

T2417-5G T2417-1G T2417-25G
TCIAT2417-5GEA 78 EUR
TCIAT2417-5G TCIAT2417-1G TCIAT2417-25G
4-(Trifluoromethyl)cyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
4-(Trifluoromethyl)cyclohexanecarboxylic acid (cis and trans mixture)
Formula: C₈H₁₁F₃O₂
MDL Number: MFCD00220049
CAS Number: 95233-30-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca6135b1771a7803639fdf5f44d9fb04c3d328c9&vid=ca6135b1771a7803639fdf5f44d9fb04c3d328c9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 % (total of isomers)
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR