Stampa…

Tetraheptylammonium bromide ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

T1602-25G T1602-5G T1602-100G
TCIAT1602-25GEA 81 EUR
TCIAT1602-25G TCIAT1602-5G TCIAT1602-100G
Tetraheptylammonium bromide ≥98.0% (by titrimetric analysis)
Tetraheptylammonium bromide
Formula: C₂₈H₆₀BrN
Punto di fusione: 96 °C
MDL Number: MFCD00011861
CAS Number: 4368-51-8
EINECS: 224-459-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=364a99e30e6eab3b7340a485aa908d0a7c5f3a3e&vid=364a99e30e6eab3b7340a485aa908d0a7c5f3a3e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 88.0 to 92.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR