Stampa…

Tetremetilammonio idrossido 10% in methanol for photoresist research

Fornitore: TCI
Pericolo

T1201-25ML
TCIAT1201-25MLEA 40 EUR
TCIAT1201-25ML
Tetremetilammonio idrossido 10% in methanol for photoresist research
Tetremetilammonio idrossido
Formula: C₄H₁₃NO
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00008280
CAS Number: 75-59-2
EINECS: 200-882-9
UN: 2924
ADR: 3,II
Merck Index: 14,09224
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5be4dc74437d365abce27792066b00c181fba48f&vid=3beb55b8a2eaa0a73ff553dfc3ccc34a830e648b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Concentration (Neutralization titration) 10.0 to 11.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR