Stampa…

2,4,6-Tris(2-pyridyl)-1,3,5-triazine ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

T0530-1G T0530-25G T0530-5G
TCIAT0530-1GEA 17 EUR
TCIAT0530-1G TCIAT0530-25G TCIAT0530-5G
2,4,6-Tris(2-pyridyl)-1,3,5-triazine ≥98.0% (by titrimetric analysis)
2,4,6-Tris(2-pyridyl)-1,3,5-triazine
Formula: C₁₈H₁₂N₆
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00006045
CAS Number: 3682-35-7
EINECS: 222-965-9
Merck Index: 14,09750
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff67a6dad76d56a73e075574ae143a6aa863da4&vid=8ff67a6dad76d56a73e075574ae143a6aa863da4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR