Stampa…

Sodium phenoxyacetate ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

S0529-25G
TCIAS0529-25GEA 122 EUR
TCIAS0529-25G
Sodium phenoxyacetate ≥98.0% (by titrimetric analysis)
Sodium phenoxyacetate
Formula: C₈H₈O₃
MDL Number: MFCD00064234
CAS Number: 3598-16-1
EINECS: 222-746-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR