Stampa…

4-Pentylphenyl 4-Methoxybenzoate ≥98.0% (by GC)

Fornitore: TCI

P2692-5G
TCIAP2692-5GEA 85 EUR
TCIAP2692-5G
4-Pentylphenyl 4-Methoxybenzoate ≥98.0% (by GC)
4-Pentylphenyl 4-Methoxybenzoate
Formula: C₁₉H₂₂O₃
Punto di fusione: 29°C
MDL Number: MFCD08276868
CAS Number: 38444-13-2
EINECS: 253-932-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR