Stampa…

Prochlorperazine maleate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

P2368-5G
TCIAP2368-5GEA 67 EUR
TCIAP2368-5G
Prochlorperazine maleate ≥98.0% (by HPLC, titration analysis)
Prochlorperazine maleate
Formula: C₂₀H₂₄ClN₃S·2C₄H₄O₄
MDL Number: MFCD00069330
CAS Number: 84-02-6
EINECS: 201-511-3
Merck Index: 14,07761
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c04e8231f1ca2971659fa98fdb9e150188ae043&vid=1c04e8231f1ca2971659fa98fdb9e150188ae043&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR