Stampa…

(Phenylthio)acetonitrile ≥98.0% (by GC)

Fornitore: TCI
Attenzione

P1602-5G
TCIAP1602-5GEA 25 EUR
TCIAP1602-5G
(Phenylthio)acetonitrile ≥98.0% (by GC)
(Phenylthio)acetonitrile
Formula: C₈H₇NS
Punto di ebollizione: 146°C
MDL Number: MFCD00019831
CAS Number: 5219-61-4
EINECS: 226-013-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9679dfc56375c7b003c3eed164f8084445a9df9f&vid=9679dfc56375c7b003c3eed164f8084445a9df9f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR