Stampa…

1,3-Propanedisulphonic acid disodium salt ≥97.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

P1205-5G
TCIAP1205-5GEA 29 EUR
TCIAP1205-5G
1,3-Propanedisulphonic acid disodium salt ≥97.0% (by titrimetric analysis)
1,3-Propanedisulphonic acid disodium salt
Formula: C₃H₆Na₂O₆S₂
MDL Number: MFCD00191483
CAS Number: 36589-58-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47515e9e1cd0161aa06253b8456d0f3cad8456c4&vid=47515e9e1cd0161aa06253b8456d0f3cad8456c4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR