Stampa…

D-(+)-Proline ≥98.0% (by titrimetric analysis)

Fornitore: TCI

Synonyms: H-D-Pro-OH, (R)-(+)-Proline, D-Proline

P0994-25G P0994-5G
TCIAP0994-25GEA 96 EUR
TCIAP0994-25G TCIAP0994-5G
D-(+)-Proline ≥98.0% (by titrimetric analysis)
D-(+)-Proline
Formula: C₅H₉NO₂
Peso molecolare: 115.13 g/mol
Punto di fusione: 223
MDL Number: MFCD00064317
CAS Number: 344-25-2
EINECS: 206-452-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5798067ab14069ccc0025d495b51a8f23978d891&vid=5798067ab14069ccc0025d495b51a8f23978d891&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +83.0 to +87.0 deg (C=4, H2O)
Solubility in Water within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR