Stampa…

PIPES sale sodico ≥99.0% (by titrimetric analysis) Good's buffer component for biological research

Fornitore: TCI

P0875-25G
TCIAP0875-25GEA 35 EUR
TCIAP0875-25G
PIPES sale sodico ≥99.0% (by titrimetric analysis) Good's buffer component for biological research
PIPES sale sodico
Formula: C₈H₁₇N₂NaO₆S₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00065472
CAS Number: 10010-67-0
EINECS: 233-005-3
Merck Index: 14,07479
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ab9fa39eee3f01694dc454bc3552edb2b4d92ea&vid=68b54802c9d91919b0b576be7b90e2c1911c0e77&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR