Stampa…

4-Piperidone monohydrate hydrochloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: 4,4-Piperidinediol Hydrochloride, 4,4-Dihydroxypiperidine Hydrochloride, 4-Ketopiperidine Monohydrate Hydrochloride, 4-Piperidinone monohydrate hydrochloride, Piperidin-4-one hydrate hydrochloride, 4-Azacyclohexanone Monohydrate Hydrochloride

P0842-500G P0842-100G
TCIAP0842-500GEA 553 EUR
TCIAP0842-500G TCIAP0842-100G
4-Piperidone monohydrate hydrochloride ≥98.0% (by titrimetric analysis)
4-Piperidone monohydrate hydrochloride
Formula: C₅H₉NO·xHCl·yH₂O
Peso molecolare: 99.13 g/mol (free base)
MDL Number: MFCD00012776
CAS Number: 40064-34-4
EINECS: 254-779-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f86ee17ed327e89f04f7a17566f7a8f4a13891e&vid=0f86ee17ed327e89f04f7a17566f7a8f4a13891e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystaline
Purity (Argentometric Titration) min. 98.0 %
Water 11.0 to 13.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR