Stampa…

Potassium tetraoxalate dihydrate ≥99.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

P0475-25G P0475-500G
TCIAP0475-25GEA 16 EUR
TCIAP0475-25G TCIAP0475-500G
Potassium tetraoxalate dihydrate ≥99.0% (by titrimetric analysis)
Potassium tetraoxalate dihydrate
Formula: C₄H₃KO₈·2H₂O
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00036364
CAS Number: 6100-20-5
EINECS: 204-874-6
UN: 2811
ADR: 6.1,III
Merck Index: 14,07689
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=890b5da9dac0bf663e34826646c59d8fc689e207&vid=890b5da9dac0bf663e34826646c59d8fc689e207&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Redox method) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR