Stampa…

Orotic acid monohydrate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

O0065-25G O0065-500G O0065-100G
TCIAO0065-25GEA 16 EUR
TCIAO0065-25G TCIAO0065-500G TCIAO0065-100G
Orotic acid monohydrate ≥98.0% (by HPLC, titration analysis)
Orotic acid monohydrate
Formula: C₅H₄N₂O₄·1H₂O
MDL Number: MFCD00149398
CAS Number: 50887-69-9
EINECS: 200-619-8
Merck Index: 14,06876
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b05f1710918dec04715f78da776bc1dbb2d4441&vid=1b05f1710918dec04715f78da776bc1dbb2d4441&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in 1mol/L NaOH almost transparency
Water 9.5 to 11.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR