Stampa…

N-Acetylneuraminic acid trimer alpha(2-8) 95,0 HPLC_ASSAY_METHOD

Fornitore: TCI

Synonyms: Neu5Ac alpha(2-8)Neu5Ac alpha(2-8)Neu5Ac

N1164-10MG
TCIAN1164-10MGEA 283 EUR
TCIAN1164-10MG
N-Acetylneuraminic acid trimer alpha(2-8) 95,0 HPLC_ASSAY_METHOD
N-Acetylneuraminic acid trimer alpha(2-8)
Formula: C₃₃H₅₃N₃O₂₅
Peso molecolare: 891.78 g/mol
Temperatura di stoccaggio: Congelamento
CAS Number: 76152-09-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR