Stampa…

N-Acetylneuraminic acid Dimer alpha(2-8) 95,0 HPLC_ASSAY_METHOD

Fornitore: TCI

Synonyms: Neu5Ac alpha(2-8)Neu5Ac

N1163-10MG
TCIAN1163-10MGEA 221 EUR
TCIAN1163-10MG
N-Acetylneuraminic acid Dimer alpha(2-8) 95,0 HPLC_ASSAY_METHOD
N-Acetylneuraminic acid Dimer alpha(2-8)
Formula: C₂₂H₃₆N₂O₁₇
Peso molecolare: 600.53 g/mol
Temperatura di stoccaggio: Congelamento
CAS Number: 95983-78-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR