Stampa…

Norlaudanosine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI
Attenzione

N0918-5G N0918-25G
TCIAN0918-5GEA 37 EUR
TCIAN0918-5G TCIAN0918-25G
Norlaudanosine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Norlaudanosine hydrochloride
Formula: C₂₀H₂₅NO₄
MDL Number: MFCD00035267
CAS Number: 6429-04-5
EINECS: 229-213-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef5debe6725e9303cb2668d7ef6063533e7045c9&vid=ef5debe6725e9303cb2668d7ef6063533e7045c9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 216.0 to 220.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR