Stampa…

(4-Nitrobenzyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

N0701-25G
TCIAN0701-25GEA 86 EUR
TCIAN0701-25G
(4-Nitrobenzyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
(4-Nitrobenzyl)triphenylphosphonium chloride
Formula: C₂₅H₂₁BrNO₂P
Punto di fusione: 275°C
MDL Number: MFCD00032108
CAS Number: 2767-70-6
EINECS: 220-444-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43ad1a24e9ab7f84aaf4344124c5aacf0668beb3&vid=43ad1a24e9ab7f84aaf4344124c5aacf0668beb3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR