Stampa…

Nonadecanoic acid methyl ester ≥98.0% (by GC)

Fornitore: TCI

N0460-25G N0460-5G
TCIAN0460-25GEA 182 EUR
TCIAN0460-25G TCIAN0460-5G
Nonadecanoic acid methyl ester ≥98.0% (by GC)
Nonadecanoic acid methyl ester
Formula: C₂₀H₄₀O₂
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00009011
CAS Number: 1731-94-8
EINECS: 217-056-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e123c8f9832315058830ffbfeb51683ed07ec2cb&vid=e123c8f9832315058830ffbfeb51683ed07ec2cb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 38.0 to 42.0 deg-C
Solubility in Ethylacetate almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR