Stampa…

3-Methyl-1,2-cyclopentanedione 98,0 GC, T

Fornitore: TCI

Synonyms: 3-Methyl-1,2-cyclopentanedione, Maple lactone, Methyl cyclopentenolone, 3-Methylcyclopentane-1,2-dione

M2904-25G
TCIAM2904-25GEA 33 EUR
TCIAM2904-25G
3-Methyl-1,2-cyclopentanedione 98,0 GC, T
3-Methyl-1,2-cyclopentanedione
Formula: C₆H₈O₂
Peso molecolare: 112.13 g/mol
Punto di ebollizione: 210
MDL Number: MFCD00001417
CAS Number: 765-70-8
EINECS: 212-154-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a03b3e848a08cda10b61179091135236ee86b119&vid=a03b3e848a08cda10b61179091135236ee86b119&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 104.0 to 108.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR