Stampa…

Fumaric acid monomethyl ester ≥98.0% (by GC)

Fornitore: TCI
Attenzione

M2413-5G M2413-25G
TCIAM2413-5GEA 53 EUR
TCIAM2413-5G TCIAM2413-25G
Fumaric acid monomethyl ester ≥98.0% (by GC)
Fumaric acid monomethyl ester
Formula: C₅H₆O₄
MDL Number: MFCD00063174
CAS Number: 2756-87-8
EINECS: 220-412-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78266193d5331d99656099d7d3cab60dc263e10e&vid=78266193d5331d99656099d7d3cab60dc263e10e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 145.0 to 149.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR