Stampa…

Gal[26Bn]β(1-4)Glc[236Bn]-β-MP ≥98.0% (by HPLC)

Fornitore: TCI

Synonyms: LacMP 3'4'-OH, per OBn, 4-Methoxyphenyl 4-O-(2,6-Di-O-benzyl-β-D-galactopyranosyl)-2,3,6-tri-O-benzyl-β-D-glucopyranoside

M1686-1G M1686-5G
TCIAM1686-1GEA 66 EUR
TCIAM1686-1G TCIAM1686-5G
Gal[26Bn]β(1-4)Glc[236Bn]-β-MP ≥98.0% (by HPLC)
Gal[26Bn]beta(1-4)Glc[236Bn]-beta-MP
Formula: C₅₄H₅₈O₁₂
Temperatura di stoccaggio: Refrigerata
CAS Number: 358681-61-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 128.0 to 132.0 deg-C
Specific rotation [a]20/D +2.0 to +5.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR