Stampa…

Sodium-2-mercapto-5-benzimidazolesulphonate dihydrate ≥98.0% (by HPLC)

Fornitore: TCI

M1270-25G
TCIAM1270-25GEA 60 EUR
TCIAM1270-25G
Sodium-2-mercapto-5-benzimidazolesulphonate dihydrate ≥98.0% (by HPLC)
Sodium-2-mercapto-5-benzimidazolesulphonate dihydrate
Formula: C₇H₆N₂O₃S₂
MDL Number: MFCD00192260
CAS Number: 207511-11-3
EINECS: 258-862-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 % (calcd.on anh.substance)
Solubility in hot Water almost transparency
Water 12.0 to 13.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR