Stampa…

Metilcellulosa, 350-550 mPa.s (20 °C) (C=2%, H₂O)

Fornitore: TCI

M0293-500G M0293-25G
TCIAM0293-500GEA 71 EUR
TCIAM0293-500G TCIAM0293-25G
Metilcellulosa, 350-550 mPa.s (20 °C) (C=2%, H₂O)
Methyl cellulose
MDL Number: MFCD00081763
CAS Number: 9004-67-5
Merck Index: 14,06040

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Drying loss max. 8.0 %
Ignition residue (Sulfate) max. 1.5 %
Viscosity 350.0 to 550.0 mPa-s (2 %, H2O, 20 deg-C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR