Stampa…

Lead(II) bromide for perovskite precursor

Fornitore: TCI
Pericolo

L0288-5G L0288-25G L0288-1G
TCIAL0288-5GEA 68 EUR
TCIAL0288-5G TCIAL0288-25G TCIAL0288-1G
Lead(II) bromide for perovskite precursor
Lead(II) bromide
Formula: Br₂Pb
Punto di fusione: 373°C
MDL Number: MFCD00011156
CAS Number: 10031-22-8
EINECS: 233-084-4
UN: 2291
ADR: 6.1,III
Merck Index: 14,05403
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0df59e07414b9a39a79fae159c6201a8e5d8fd7a&vid=0df59e07414b9a39a79fae159c6201a8e5d8fd7a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR