Stampa…
2-Chloroethyl laurate ≥95.0% (by GC)
Fornitore: TCI
Synonyms: 2-Chloroethyl dodecanoate, Lauric Acid 2-Chloroethyl Ester
L0106-25G
TCIAL0106-25GEA 468 EUR
TCIAL0106-25G
2-Chloroethyl laurate ≥95.0% (by GC)
2-Chloroethyl laurate
Formula: C₁₄H₂₇ClO₂
Peso molecolare: 262.82 g/mol
Punto di ebollizione: 187°C
MDL Number: MFCD00059475
CAS Number: 64919-15-9
Visualizza i codici
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %