Stampa…

Calcium levulinate dihydrate ≥98.0% (by titrimetric analysis)

Fornitore: TCI

L0043-25G
TCIAL0043-25GEA 92 EUR
TCIAL0043-25G
Calcium levulinate dihydrate ≥98.0% (by titrimetric analysis)
Calcium levulinate dihydrate
Formula: C₅H₈O₃
MDL Number: MFCD00045959
CAS Number: 5743-49-7
EINECS: 209-725-9
Merck Index: 14,01679

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Water 10.0 to 13.0 %
Melting point 121.0 to 125.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR