Stampa…

Levulinic acid ≥97.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

L0042-25G L0042-500G
TCIAL0042-25GEA 17 EUR
TCIAL0042-25G TCIAL0042-500G
Levulinic acid ≥97.0% (by GC, titration analysis)
Levulinic acid
Formula: C₅H₈O₃
Punto di ebollizione: 245 °C (1013 hPa)
Punto di fusione: 26…29 °C
Densità: 1,14 g/cm³ (20 °C)
Flash Pt: 138 °C
MDL Number: MFCD00002796
CAS Number: 123-76-2
EINECS: 204-649-2
UN: 3261
ADR: 8,III
Merck Index: 14,05472

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR