Stampa…

Isobutyryl bromide ≥98.0% (by GC)

Fornitore: TCI
Pericolo

I0114-25G I0114-5G
TCIAI0114-25GEA 204 EUR
TCIAI0114-25G TCIAI0114-5G
Isobutyryl bromide ≥98.0% (by GC)
Isobutyryl bromide
Formula: C₄H₇BrO
Peso molecolare: 151.00 g/mol
Punto di ebollizione: 117°C
MDL Number: MFCD00059468
CAS Number: 2736-37-0
UN: 2920
ADR: 8 II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR