Stampa…

Imidazolo ≥99.8% (by GC), zone refined: number of passes 30

Fornitore: TCI

Synonyms: 1H-Imidazolo

I0014-1SAMPLE
TCIAI0014-1SAMPLEEA 137 EUR
TCIAI0014-1SAMPLE
Imidazolo ≥99.8% (by GC), zone refined: number of passes 30
Imidazolo

CAS Number: 288-32-4
MDL Number: MFCD00005183
Molecular Formula: C3H4N2
Molecular Weight: 68.08
Purity/Analysis Method: >99.8% (GC)
Form: Crystal
Melting point (°C): 92
Flash Point (°C): 145

Formula: C₃H₄N₂
Peso molecolare: 68.08 g/mol
Punto di ebollizione: 255
Punto di fusione: 92 °C
Flash Pt: 145 °C
MDL Number: MFCD00005183
CAS Number: 288-32-4
UN: 2923
ADR: 8 III
Merck Index: 13,04935

Visualizza i codici

Risultati del test di specificazione

Purity/Analysis Method >99.8% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR