Stampa…

cis-4-(Hydroxymethyl)cyclohexanecarboxylic acid ≥98.0% (by GC)

Fornitore: TCI
Attenzione

H1242-25G H1242-5G H1242-1G
TCIAH1242-25GEA 531 EUR
TCIAH1242-25G TCIAH1242-5G TCIAH1242-1G
cis-4-(Hydroxymethyl)cyclohexanecarboxylic acid ≥98.0% (by GC)
cis-4-(Hydroxymethyl)cyclohexanecarboxylic acid
Formula: C₈H₁₄O₃
CAS Number: 73094-35-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 117.0 to 121.0 deg-C
Solubility in Methanol almost transparency

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR