Stampa…

4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) ≥98.0% (by GC) free radical

Fornitore: TCI
Attenzione

H0865-25G H0865-5G
TCIAH0865-25GEA 205 EUR
TCIAH0865-25G TCIAH0865-5G
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) ≥98.0% (by GC) free radical
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL)
Formula: C₉H₁₈NO₂
MDL Number: MFCD00006478
CAS Number: 2226-96-2
EINECS: 218-760-9
Merck Index: 14,09141
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99afd5985eaf7e307427bf134243c7342cdc93df&vid=08c5a744115963e43cfac7de06d55920c4d4831c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 97.0 %
Melting point 69.0 to 75.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR