Stampa…

Geraniol ≥96.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: (E)-3,7-Dimethylocta-2,6-dien-1-ol

G0027-25ML G0027-500ML G0027-100ML
TCIAG0027-25MLEA 16 EUR
TCIAG0027-25ML TCIAG0027-500ML TCIAG0027-100ML
Geraniol ≥96.0% (by GC)
Geraniol
Formula: C₁₀H₁₈O
Peso molecolare: 154.25 g/mol
Flash Pt: 112°C
MDL Number: MFCD00002917
CAS Number: 106-24-1
EINECS: 203-377-1
Merck Index: 14,04403
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb7e8a01e1abdd5ba4d441c44c806a4bc8d9a898&vid=bb7e8a01e1abdd5ba4d441c44c806a4bc8d9a898&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR