Stampa…

4-Fluoro-L-phenylalanine ≥98.0% (by titrimetric analysis)

Fornitore: TCI

F0274-100MG
TCIAF0274-100MGEA 36 EUR
TCIAF0274-100MG
4-Fluoro-L-phenylalanine ≥98.0% (by titrimetric analysis)
4-Fluoro-L-phenylalanine
Formula: C₉H₁₀FNO₂
MDL Number: MFCD00063064
CAS Number: 1132-68-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f3009e94acc39afeba2b4fbdd25ef3a1f3a0cdb&vid=9f3009e94acc39afeba2b4fbdd25ef3a1f3a0cdb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -23.0 to -27.0 deg (C=1,H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR