Stampa…

Disodium fumarate ≥98.0% (by titrimetric analysis)

Fornitore: TCI

F0070-25G F0070-500G
TCIAF0070-25GEA 16 EUR
TCIAF0070-25G TCIAF0070-500G
Disodium fumarate ≥98.0% (by titrimetric analysis)
Disodium fumarate
Formula: C₄H₂Na₂O₄
MDL Number: MFCD00064567
CAS Number: 17013-01-3
EINECS: 241-087-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73c54659eabc31e140c089d45a155d4c3b6df601&vid=73c54659eabc31e140c089d45a155d4c3b6df601&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR