Stampa…

Enalapril maleate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI

E1010-1G E1010-5G
TCIAE1010-1GEA 40 EUR
TCIAE1010-1G TCIAE1010-5G
Enalapril maleate ≥98.0% (by HPLC, titration analysis)
Enalapril maleate
Formula: C₂₀H₂₈N₂O₅
MDL Number: MFCD00133304
CAS Number: 76095-16-4
EINECS: 278-375-7
Merck Index: 14,03567

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -40.0 to -43.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR