Stampa…

4,5-Dihydro-2,5-dimethyl-4-oxofuran-3-yl butyrate 96,0 GC_ASSAY_METHOD

Fornitore: TCI

Synonyms: 4-Butyryloxy-2,5-dimethyl-3(2H)-furanone, Butyric Acid 4,5-Dihydro-2,5-dimethyl-4-oxofuran-3-yl

D4451-25G
TCIAD4451-25GEA 53 EUR
TCIAD4451-25G
4,5-Dihydro-2,5-dimethyl-4-oxofuran-3-yl butyrate 96,0 GC_ASSAY_METHOD
4,5-Dihydro-2,5-dimethyl-4-oxofuran-3-yl butyrate
Formula: C₁₀H₁₄O₄
Peso molecolare: 198.22 g/mol
MDL Number: MFCD08457803
CAS Number: 114099-96-6

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR