Stampa…

2,5-Dimethyl-3-furancarboxylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

D3863-5G
TCIAD3863-5GEA 174 EUR
TCIAD3863-5G
2,5-Dimethyl-3-furancarboxylic acid ≥98.0% (by GC, titration analysis)
2,5-Dimethyl-3-furancarboxylic acid
Formula: C₇H₈O₃
MDL Number: MFCD00082181
CAS Number: 636-44-2
EINECS: 211-257-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystaline
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 135.0 to 139.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR