Stampa…

2-Chloro-5-methyl-1,4-phenylenediamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 2-Chloro-5-methylbenzene-1,4-diamine, 2,5-Diamino-4-chlorotoluene

D2179-25G
TCIAD2179-25GEA 155 EUR
TCIAD2179-25G
2-Chloro-5-methyl-1,4-phenylenediamine ≥98.0% (by GC, titration analysis)
2-Chloro-5-methyl-1,4-phenylenediamine
Formula: C₇H₉ClN₂
Peso molecolare: 156.61 g/mol
MDL Number: MFCD00190684
CAS Number: 5307-03-9
EINECS: 226-162-4

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 146.0 to 149.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR