Stampa…

Destrano, M.W. ~ 70000 (Dextran 70)

Fornitore: TCI

D1449-500G D1449-25G D1449-100G
TCIAD1449-500GEA 345 EUR
TCIAD1449-500G TCIAD1449-25G TCIAD1449-100G
Destrano, M.W. ~ 70000 (Dextran 70)
Destrano
Formula: (C₆H₁₀O₅)n
MDL Number: MFCD00130935
CAS Number: 9004-54-0
EINECS: 232-677-5
Merck Index: 14,02948

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline
Specific rotation [a]20/D +170 to +200 deg (C=6, H2O)
pH 5.0 to 7.0 (60g/L, 25deg-C)
limiting viscosity 0.21 to 0.26 cp


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR