Stampa…

3-(3,4-Dihydroxyphenyl)-DL-alanine ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

D0599-1G
TCIAD0599-1GEA 51 EUR
TCIAD0599-1G
3-(3,4-Dihydroxyphenyl)-DL-alanine ≥98.0% (by titrimetric analysis)
3-(3,4-Dihydroxyphenyl)-DL-alanine
Formula: C₉H₁₁NO₄
MDL Number: MFCD00063060
CAS Number: 63-84-3
EINECS: 200-566-0
Merck Index: 14,03420
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efd6120669451c7f9472dd654819d5191f9e332a&vid=efd6120669451c7f9472dd654819d5191f9e332a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in 1mol/L HCl almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR