Stampa…

Calcium cyanamide

Fornitore: TCI
Pericolo

C0006-25G C0006-500G
TCIAC0006-25GEA 16 EUR
TCIAC0006-25G TCIAC0006-500G
Calcium cyanamide
Calcium cyanamide
Formula: CCaN₂
MDL Number: MFCD00064894
CAS Number: 156-62-7
EINECS: 205-861-8
UN: 1403
ADR: 4.3,III
Merck Index: 14,01662
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a8f432aac00270d483292a9b66e56feec53321c&vid=8a8f432aac00270d483292a9b66e56feec53321c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light gray to Brown to Black powder to crystal
Nitorogen of cyanamide min. 19.5 %

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR