Stampa…
SDS-PAGE Sample Buffer, 2X, (2-Mercaptoethanol-free) for Electrophoresis
Fornitore: TCI
B5834-25ML
TCIAB5834-25MLEA 54 EUR
TCIAB5834-25ML
SDS-PAGE Sample Buffer, 2X, (2-Mercaptoethanol-free) for Electrophoresis
Soluzioni tampone Tamponi per elettroforesi Tamponi per gel loading
SDS-PAGE Sample Buffer is specially formulated for protein sample preparation to based on the Laemmli Method of SDS-PAGE system.
This product contains 4% SDS, 20% glycerol, 0,01% bromophenol blue and 0,125 M Tris HCl, pH approx. 6,8. Does not contain 2-Mercaptoethanol.
Attenzione: Please use after bringing back to normal temperature.