Stampa…

1-Boc-3-fluoro-4-piperidone ≥97.0% (by GC)

Fornitore: TCI
Attenzione

B4858-200MG B4858-1G
TCIAB4858-200MGEA 76 EUR
TCIAB4858-200MG TCIAB4858-1G
1-Boc-3-fluoro-4-piperidone ≥97.0% (by GC)
1-Boc-3-fluoro-4-piperidone
Formula: C₁₀H₁₆FNO₃
MDL Number: MFCD10566554
CAS Number: 211108-50-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8de80e375979a577d85884a1b61b890c4154f2a0&vid=8de80e375979a577d85884a1b61b890c4154f2a0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 74.0 to 78.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR