Stampa…

Boc-L-norvaline ≥98.0% (by titrimetric analysis)

Fornitore: TCI

B4320-1G B4320-5G
TCIAB4320-1GEA 20 EUR
TCIAB4320-1G TCIAB4320-5G
Boc-L-norvaline ≥98.0% (by titrimetric analysis)
Boc-L-norvaline
Formula: C₁₀H₁₉NO₄
MDL Number: MFCD00037268
CAS Number: 53308-95-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7ed7576e4b82f5165dec567f31116b2b0ce341d&vid=5f14b171e219bee9a36ebdd0d870051b8584de38&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 64.0 to 69.0 deg-C
Specific rotation [a]20/D -14.0 to -17.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR