Stampa…

N-(tert-Butoxycarbonyl)-1,4-phenylenediamine ≥95.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

B4081-5G B4081-25G
TCIAB4081-5GEA 70 EUR
TCIAB4081-5G TCIAB4081-25G
N-(tert-Butoxycarbonyl)-1,4-phenylenediamine ≥95.0% (by HPLC, titration analysis)
N-(tert-Butoxycarbonyl)-1,4-phenylenediamine
Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD00043022
CAS Number: 71026-66-9
EINECS: 275-132-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 114.0 to 118.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR