Stampa…

Butil 4-idrossibenzoato ≥99.0% (by GC, titration analysis) for biochemical research

Fornitore: TCI
Attenzione

B3771-25G B3771-5G
TCIAB3771-25GEA 26 EUR
TCIAB3771-25G TCIAB3771-5G
Butil 4-idrossibenzoato ≥99.0% (by GC, titration analysis) for biochemical research
Butil 4-idrossibenzoato
Formula: C₁₁H₁₄O₃
Punto di ebollizione: 156°C
Punto di fusione: 73 °C
Densità: 1,23 g/cm³ (20 °C)
MDL Number: MFCD00016478
CAS Number: 94-26-8
EINECS: 202-318-7
Merck Index: 14,01584
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29bbc8d48629d1c2a3a77c48cb28fc923d3f199f&vid=1f2588ec19e5b3fd99566c94b3c2486059bb1183&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %
Melting point 69.0 to 72.0 deg-C
Protein impurity test (SDS-PAGE) to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR