Stampa…

Phenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI
Attenzione

B0857-25G B0857-250G B0857-5G
TCIAB0857-25GEA 27 EUR
TCIAB0857-25G TCIAB0857-250G TCIAB0857-5G
Phenylboronic acid (contains varying amounts of Anhydride)
Phenylboronic acid
Formula: C₆H₇BO₂
Punto di fusione: 217…220 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002103
CAS Number: 98-80-6
EINECS: 202-701-9
Merck Index: 14,01068
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=734d53715068835d4d746ed2356dc5838b77763b&vid=734d53715068835d4d746ed2356dc5838b77763b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Dark green powder to crystal
Purity (Neutralization titration) 97.0 to 117.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR