Stampa…

1,4-Bis(4-methyl-5-phenyloxazol-2-yl)benzene ≥99.0% (by HPLC, total nitrogen) for scintillation spectrometry

Fornitore: TCI

B0499-25G
TCIAB0499-25GEA 330 EUR
TCIAB0499-25G
1,4-Bis(4-methyl-5-phenyloxazol-2-yl)benzene ≥99.0% (by HPLC, total nitrogen) for scintillation spectrometry
1,4-Bis(4-methyl-5-phenyloxazol-2-yl)benzene
Formula: C₂₆H₂₀N₂O₂
Punto di fusione: 234…235 °C
MDL Number: MFCD00005310
CAS Number: 3073-87-8
EINECS: 221-349-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad335372f20f4079013715510186bdd02508c983&vid=024612035e12ac077f48ca797814a5ccaa5659bb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Green powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (with Total Nitrogen) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR