Stampa…

1,4-Benzoquinone dioxime 95,0 N_ASSAY_METHOD

Fornitore: TCI
Attenzione

Synonyms: p-Quinone Dioxime

B0090-25G
TCIAB0090-25GEA 16 EUR
TCIAB0090-25G
1,4-Benzoquinone dioxime 95,0 N_ASSAY_METHOD
1,4-Benzoquinone dioxime
Formula: C₆H₆N₂O₂
Peso molecolare: 138.13 g/mol
MDL Number: MFCD00063636
CAS Number: 105-11-3
EINECS: 203-271-5

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (with Total Nitrogen) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR